missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ HELIOS Polyclonal Antibody

Beskrivning
Keep as concentrated solution. Predicted reactivity: Mouse (99%), Rat (99%), Rhesus Monkey (100%). Positive Control: Jurkat, Jurkat NE. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the Ikaros family of zinc-finger proteins. Three members of this protein family (Ikaros, Aiolos and Helios) are hematopoietic-specific transcription factors involved in the regulation of lymphocyte development. This protein forms homo- or hetero-dimers with other Ikaros family members, and is thought to function predominantly in early hematopoietic development. Multiple transcript variants encoding different isoforms have been found for this gene, but the biological validity of some variants has not been determined.
Specifikationer
Specifikationer
| Antigen | HELIOS |
| Användningsområden | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot |
| Klassificering | Polyclonal |
| Koncentration | 1 mg/mL |
| Konjugera | Unconjugated |
| Formulering | PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 |
| Gen | IKZF2 |
| Genaccessionsnr. | P81183, Q9UKS7 |
| Gene Alias | A730095J18Rik; ANF1A2; Helios; IKAROS family zinc finger 2; IKAROS family zinc finger 2 (Helios); ikaros family zinc finger protein 2; Ikzf2; Zfpn1a2; zinc finger DNA binding protein Helios; zinc finger protein Helios; zinc finger protein, subfamily 1A, 2 (Helios); ZNF1A2; Znfn1a2 |
| Gensymboler | IKZF2 |
| Visa mer |
Name des Produkts
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.
Haben Sie Verbesserungsvorschläge?