Läs mer
Epredia™ Hematology Phosphate Buffer
Produktkod. 12530037 Handla allt Epredia Produkter
Change view
pH:
6,6
7,2
Kvantitet:
9,25 g
12 x 9,25 g
Förpackningsstorlek:
Paket med 12
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. pH Kvantitet Förpackningsstorlek
12530037 7,2 12 x 9,25 g Paket med 12
12520037 6,6 9,25 g Paket med 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 12530037 Leverantör Epredia™ Leverantörsnummer 89033
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Combine Epredia™ Phosphate Buffer with Giemsa, Wright, and Wright-Giemsa stains for excellent results.
Supplied as 9.25g powder.
TRUSTED_SUSTAINABILITY
Specifikationer
Produkttyp Phosphate Buffered Saline (PBS)
Kvantitet 12 x 9,25 g
pH 7,2