Läs mer
Heme Oxygenase-1-IN-1 hydrochloride, MedChemExpress
Produktkod. 16465386
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16465386 100 mg -
16450837 50 mg -
16447247 5 mg -
16414538 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16465386 Leverantör Medchem Express Leverantörsnummer HY111798A
MedChemExpress Heme Oxygenase-1-IN-1 (Compound 2) hydrochloride is a heme oxygenase 1 (HO-1) inhibitor with an IC50 of 0.25 μM.
Heme Oxygenase-1-IN-1 (Compound 2) hydrochloride is a heme oxygenase 1 (HO-1) inhibitor with an IC50 of 0.25 μM.
Specifikationer
Kemiskt namn eller material Heme Oxygenase-1-IN-1 hydrochloride
CAS 1092851-70-1
Färg Light Yellow
Anteckningar om renhetsgrad Research
Molekylformel C13H16BrClN2
Kvantitet 100 mg
Löslighetsinformation DMSO : 125 mg/mL (396.02 mM; Need ultrasonic) ∣H2O : 100 mg/mL (316.82 mM; Need ultrasonic)
LEDER BrC1=CC=C(CCCCN2C=CN=C2)C=C1.Cl
Molekylvikt (g/mol) 315.64
Formel vikt 315.64
Procent renhet 99.03%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death