missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Hemoglobin, Human, Lyoph.
Brand: MP Biomedicals™ 0855914
Additional Details : Weight : 0.00010kg
Specifications
22mg/mL | |
Dialyzed into 0.01M Sodium Phosphate, 0.07M Sodium Chloride, pH 7.3, filtered through a 0.22μm filter, vialed and lyophilized | |
7.3 | |
Gel Filtration/Ion-Exchange Chromatography, Immunoabsorption, Salt Fractionation | |
Human Red Blood Cells | |
Lyophilized |
Immunoassay | |
Hemoglobin from Human red blood cells | |
Burette Procedure | |
20 mg | |
2° to 8°C |