Learn More
BINDER™ HEPA Air Filter
BINDER™ HEPA Air Filter is an accessory for unit FED 400 Drying and Heating Chamber.
Brand: BINDER™ 8012-0076
17708.00 SEK valid until 2024-04-30
Use promo code "21656" to get your promotional price.
Additional Details : Weight : 1.00000kg
Description
- On fresh air supply for unit; filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% at 0.3 μm)
Order Info
ForUseWithApplication
Specifications
HEPA air filter - On fresh air supply for unit; filter classification H14 (acc. to. EN 1822-1:2009, > 99.995% (at 0.3 μm)) | |
BINDER Chambers FED 400 | |
HEPA Filter |