Läs mer
Heparan Sulfate (Technical Grade), TRC
Produktkod. 30134192
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30134192 10 mg -
30133242 2.5 mg 2.5mg
30012522 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30134192 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH24578010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 9050-30-0
Molekylformel C26H45N3O39S6
Molekylvikt (g/mol) 1216
Synonym Alpha-idosane, HHS 5, Heparan N-Sulfate, Heparan Sulfate, Heparan Sulphate, Heparitin, Heparitin Monosulfate, N-Acetylheparan Sulfate, Suleparoid, Tavidan;
Specifikationer
Kemiskt namn eller material Heparan Sulfate