missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heparin, MP Biomedicals™
Click to view available options
Quantity:
10 mg
50 mg
100 mg
250 mg
Specifications
Color | Colorless or White |
Quantity | 10 mg |
Formula Weight | ≈3000 |
Physical Form | Powder or Solution |
Chemical Name or Material | Heparin, Sodium Salt from Porcine |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- Heparin Sodium Salt, Porcine
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P330-Rinse mouth.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : heparin sodium salt
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'