missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
HepcidinAntimicrobialPeptide Rabbit anti-Human, Clone: 1033F, R&D Systems™

Rabbit Monoclonal Antibody
Brand: R&D Systems MAB83071-SP
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Hepcidin Antimicrobial Peptide Monoclonal specifically detects Hepcidin Antimicrobial Peptide in Human samples. It is validated for ELISA.Specifikationer
| HepcidinAntimicrobialPeptide | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2μm filtered solution in PBS. | |
| HAMP, HEPC, hepcidin antimicrobial peptide, HEPCPutative liver tumor regressor, HFE2B, HFE2Bhepcidin, LEAP1, LEAP-1, LEAP1PLTR, Liver-expressed antimicrobial peptide 1, PLTR | |
| Synthetic peptide containing human Hepcidin Accession # P81172 | |
| 25 μg | |
| 57817 | |
| Reconstitute at 0.5mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
| ELISA | |
| 1033F | |
| ELISA | |
| P81172 | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detect human Hepcidin in direct ELISAs. | |
| Human | |
| Supernatant |
Produkttitel
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.
Hittar du en möjlighet till förbättring?