missing translation for 'onlineSavingsMsg'
Learn More

HEPES sodium salt, >98%, MP Biomedicals™

Catalog Number p-4404609
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
This item is not returnable. View return policy
75277-39-3
C8H17N2NaO4S
260.28
MFCD00036463
RDZTWEVXRGYCFV-UHFFFAOYSA-M
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt
2724248
CHEBI:46758
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
This item is not returnable. View return policy
75277-39-3
C8H17N2NaO4S
260.28
MFCD00036463
RDZTWEVXRGYCFV-UHFFFAOYSA-M
4-(2-Hydroxyethyl)piperazine-1-ethanesulfonic acid sodium salt, N-(2-Hydroxyethyl)piperazine-N'-(2-ethanesulfonic acid) sodium salt
2724248
CHEBI:46758
sodium 2-[4-(2-hydroxyethyl)piperazin-1-yl]ethane-1-sulfonate
[Na+].OCCN1CCN(CCS([O-])(=O)=O)CC1
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range >98%
Formula Weight 260.3
Chemical Name or Material HEPES sodium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'