Läs mer
Heptadecanoic Acid 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl Ester, TRC
Produktkod. 17952975
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17952975 25 mg
17922575 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17952975 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH28104525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1404053-62-8
Molekylformel C40 H59 F N4 O4
Molekylvikt (g/mol) 678.92
Synonym Heptadecanoic acid, 3-[2-[4-(6-fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl ester, 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)-1-piperidinyl]ethyl]-6,7,8,9-tetrahydro-2-methyl-4-oxo-4H-pyrido[1,2-a]pyrimidin-9-yl heptadecanoate, 9-Hydroxyrisperidone Heptadecanoate, Paliperidone Margarate, 3-[2-[4-(6-Fluoro-1,2-benzisoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-4-oxo-6,7,8,9-tetrahydro-4H-pyrido[1,2-a]pyrimidin-9-yl Heptadecanoate
IUPAC-namn [3-[2-[4-(6-fluoro-1,2-benzoxazol-3-yl)piperidin-1-yl]ethyl]-2-methyl-4-oxo-6,7,8,9-tetrahydropyrido[1,2-a]pyrimidin-9-yl] heptadecanoate
LEDER CCCCCCCCCCCCCCCCC(=O)OC1CCCN2C(=O)C(=C(C)N=C12)CCN3CCC(CC3)c4noc5cc(F)ccc45
Specifikationer
Kemiskt namn eller material Paliperidone Heptadecanoate
Kvantitet 25 mg
InChI formel InChI=1S/C40H59FN4O4/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-19-37(46)48-35-18-17-25-45-39(35)42-30(2)33(40(45)47)24-28-44-26-22-31(23-27-44)38-34-21-20-32(41)29-36(34)49-43-38/h20-21,29,31,35H,3-19,22-28H2,1-2H3
Formel vikt 678.452