Läs mer
Heptadecanoyl Chloride, TRC
Produktkod. 30407374
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30407374 100 mg
1 options
Produktkod. 30407374 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH281055100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Heptadecanoyl Chloride
CAS 40480-10-2
Molekylformel C17H33ClO
Kvantitet 100 mg
Synonym Heptadecanoic Acid Chloride, n-Heptadecanoyl Chloride
InChI formel InChI=1S/C17H33ClO/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19/h2-16H2,1H3
LEDER ClC(CCCCCCCCCCCCCCCC)=O
IUPAC-namn heptadecanoyl chloride
Molekylvikt (g/mol) 288.9
Formel vikt 288.22