Läs mer
Heptaethylene Glycol Monododecyl Ether, TRC
Produktkod. 30137810
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137810 1 g
30133317 500 mg
17945065 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137810 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2802851G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 3055-97-8
Molekylformel C26H54O8
Molekylvikt (g/mol) 494.7
Synonym AEO 7, C12E7, C12EO7, Dodecanol Heptaethoxy Ether, 2-[2-[2-[2-[2-[2-[2-(Dodecyloxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]-ethanolHeptaethylene Glycol Dodecyl Ether, Heptaethylene Glycol n-Dodecyl Ether, Heptaethylene Glycol, Monododecyl ether, Heptaethyleneglycol monolauryl Ether, Mergital LT 7, Nikkol BL 7SY, Pingpingjia 7
IUPAC-namn 2-[2-[2-[2-[2-[2-(2-dodecoxyethoxy)ethoxy]ethoxy]ethoxy]ethoxy]ethoxy]ethanol
LEDER CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCO
Specifikationer
Kemiskt namn eller material Heptaethylene Glycol Monododecyl Ether
Kvantitet 1 g
InChI formel InChI=1S/C26H54O8/c1-2-3-4-5-6-7-8-9-10-11-13-28-15-17-30-19-21-32-23-25-34-26-24-33-22-20-31-18-16-29-14-12-27/h27H,2-26H2,1H3
Formel vikt 494.3819