missing translation for 'onlineSavingsMsg'
Läs mer

Heptanenitrile, 98%, Thermo Scientific Chemicals

Produktkod. p-7082071
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 g
25 g
Förpackningsstorlek:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet Förpackningsstorlek
11409443 5 g 5g
11419443 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11409443 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer A12326.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 629-08-3 | Molecular Formula: C7H13N | Molecular Weight: 111.19 g/mol

The Heptanenitrile is used for producing 2, 3, 5, 6-tetrahexyl-pyrazine. As intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The Heptanenitrile is used for producing 2, 3, 5, 6-tetrahexyl-pyrazine. As intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, Strong reducing agents, Strong acids, Strong bases.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 629-08-3
Molekylformel C7H13N
Molekylvikt (g/mol) 111.19
MDL-nummer MFCD00039503
InChI-nyckel SDAXRHHPNYTELL-UHFFFAOYSA-N
Synonym heptanonitrile, hexyl cyanide, enanthonitrile, 1-cyanohexane, 1-heptanonitrile, heptane-1-nitrile, n-heptanenitrile, unii-ua4z3qt98a, ua4z3qt98a, heptane nitrile
PubChem CID 12372
IUPAC-namn heptanenitrile
LEDER CCCCCCC#N

Specifikationer

Kemiskt namn eller material Heptanenitrile
Smältpunkt -64°C
Kokpunkt 186°C to 187°C
Flampunkt 58°C (136°F)
Kvantitet 5 g
UN-nummer UN1992
Beilstein 1740405
Formel vikt 111.19
Procent renhet 98%

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.