missing translation for 'onlineSavingsMsg'
Läs mer

Heptyl Undecyl Phthalate, TRC

Produktkod. 30138028
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138028 250 mg
30012045 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30138028 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH285605250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65185-88-8
Molekylformel C26H42O4
Molekylvikt (g/mol) 418.61
Synonym 1,2-Benzenedicarboxylic Acid Heptyl Undecyl Ester, 1,2-Benzenedicarboxylic Acid 1-Heptyl 2-Undecyl Ester
IUPAC-namn 1-O-heptyl 2-O-undecyl benzene-1,2-dicarboxylate
LEDER CCCCCCCCCCCOC(=O)c1ccccc1C(=O)OCCCCCCC

Specifications

Kemiskt namn eller material Heptyl Undecyl Phthalate
Kvantitet 250 mg
InChI formel InChI=1S/C26H42O4/c1-3-5-7-9-10-11-12-14-18-22-30-26(28)24-20-16-15-19-23(24)25(27)29-21-17-13-8-6-4-2/h15-16,19-20H,3-14,17-18,21-22H2,1-2H3
Formel vikt 418.3083
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.