Learn More
Herpes Simplex Virus 1 Antibody (HSVI/8375R) - BSA Free, R&D Systems™
Product Code. 18609743 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
Product Code. Quantity Unit Size
18609743 100 μg 100µg
1 options
Product Code. 18609743 Supplier Novus Biologicals Supplier No. NBP320741
 Restricted Item
Beräknad leverans: 27-11-2026
A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody
Herpes Simplex Virus 1 Monoclonal antibody specifically detects Herpes Simplex Virus 1 in Virus samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Antigen Herpes Simplex Virus 1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone HSVI/8375R
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL
Formulation 10mM PBS
Gene Alias Herpes simplex virus, Herpes simplex virus 1, Herpes simplex virus type 1, HSV 1, Human herpesvirus 1, Human herpesvirus type 1
Host Species Rabbit
Immunogen Detergent-solubilized herpes simplex virus (HSV) type 1 infected cells
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at -20°C to -70°C. Avoid freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG κ
Product Line Novus