Läs mer
Hexachlorobenzene-13C6, TRC
Produktkod. 30007515
Change view
Kvantitet:
1 mg
10 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30007515 1 mg
30006997 10 mg
30005414 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30007515 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2908171MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 93952-14-8
Molekylformel 13C6 Cl6
Molekylvikt (g/mol) 290.74
Synonym Benzene-13C6, hexachloro- (9CI), Hexachlorobenzene 13C6
IUPAC-namn 1,2,3,4,5,6-hexachloro(1,2,3,4,5,6-^{13}C_{6})cyclohexa-1,3,5-triene
LEDER Cl[13c]1[13c](Cl)[13c](Cl)[13c](Cl)[13c](Cl)[13c]1Cl
Specifikationer
Kemiskt namn eller material Hexachlorobenzene 13C6
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/i1+1,2+1,3+1,4+1,5+1,6+1
Formel vikt 287.8332
Procent renhet >95%