Läs mer
Hexachlorobicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic Acid, TRC
Produktkod. 30137531
Change view
Kvantitet:
1 g
2.5 g
5 g
Förpackningsstorlek:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30137531 2.5 g 2.5g
30137494 5 g -
30012486 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137531 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2907682.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 115-28-6
Molekylformel C9 H4 Cl6 O4
Molekylvikt (g/mol) 388.84
Synonym Chlorendic acid, 5-Norbornene-2,3-dicarboxylic acid, 1,4,5,6,7,7-hexachloro- (6CI,8CI), 1,4,5,6,7,7-Hexachlorobicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid, 1,4,5,6,7,7-Hexachloro-5-norbornene-2,3-dicarboxylic acid, 1,4,5,6,7,7-Hexachlorobicyclo-(2,2,1)hept-5-en-2,3-dicarboxylic acid, 1,4,5,6,7,7-Hexachlorodicyclo-(2.2.1)-5-heptene-2,3-dicarboxylic acid, 2H,3H-Hexachlorobicyclo[2.2.1]hept-5-ene-2,3-dicarboxylic acid, Chlorendic acid, HET acid, Hexachloroendomethylenetetrahydrophthalic acid, NSC 22231, NSC 41876
IUPAC-namn (1S,4R)-1,2,3,4,7,7-hexachlorobicyclo[2.2.1]hept-2-ene-5,6-dicarboxylic acid
LEDER OC(=O)C1C(C(=O)O)[C@]2(Cl)C(=C(Cl)[C@@]1(Cl)C2(Cl)Cl)Cl
Specifikationer
Kemiskt namn eller material Chlorendic acid
Kvantitet 2.5 g
InChI formel InChI=1S/C9H4Cl6O4/c10-3-4(11)8(13)2(6(18)19)1(5(16)17)7(3,12)9(8,14)15/h1-2H,(H,16,17)(H,18,19)/t1?,2?,7-,8+
Formel vikt 385.8241