Läs mer
Hexaconazole, TRC
Produktkod. 30403723
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30403723 250 mg
1 options
Produktkod. 30403723 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH293555250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Hexaconazole
CAS 79983-71-4
Anteckningar om renhetsgrad HPLC
Molekylformel C14 H17 Cl2 N3 O
Kvantitet 250 mg
Synonym 1H-1,2,4-Triazole-1-ethanol, α-butyl-α-(2,4-dichlorophenyl)-, (±)-, α-Butyl-α-(2,4-dichlorophenyl)-1H-1,2,4-triazole-1-ethanol, (±)-Hexaconazole, 2-(2,4-Dichlorophenyl)-1-(1H-1,2,4-triazol-1-yl)hexan-2-ol, Anvil, Anvil L, Anvil Liquid, Blin Exa, Canvil, Clortriafol, Contaf, Contaf 5EC, Contaf Plus, Controll, FD 4053, Flowmax 5SC, Hexaconazole, Hexaconazole Technical, Hexadhan, PC 1002, PP 523, R 154523, RS-2-(2,4-Dichlorophenyl)-1-(1H-1,2,4-triazol-1-yl)hexan-2-ol, Ranvil, Samarth, Sitara, TDP, 1H-1,2,4-Triazole-1-ethanol, α-butyl-α-(2,4-dichlorophenyl)-
InChI formel InChI=1S/C14H17Cl2N3O/c1-2-3-6-14(20,8-19-10-17-9-18-19)12-5-4-11(15)7-13(12)16/h4-5,7,9-10,20H,2-3,6,8H2,1H3
LEDER CCCCC(O)(Cn1cncn1)c2ccc(Cl)cc2Cl
IUPAC-namn 2-(2,4-dichlorophenyl)-1-(1,2,4-triazol-1-yl)hexan-2-ol
Molekylvikt (g/mol) 314.21
Formel vikt 313.0749