Läs mer
Hexacosanoic Acid ((1S,2S,3R)-2,3-Dihydroxy-1-hydroxymethyl-heptadecyl)-amide, TRC
Produktkod. 30406007
Change view
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30406007 10 mg
30392806 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30406007 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH29100810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Hexacosanoic Acid ((1S,2S,3R)-2,3-Dihydroxy-1-hydroxymethyl-heptadecyl)-amide
CAS 182362-51-2
Molekylformel C44 H89 N O4
Kvantitet 10 mg
Synonym [1S-(1R*,2R*,3S*)]-N-[2,3-dihydroxy-1-(hydroxymethyl)heptadecyl]-Hexacosanamide, N-[(1S,2S,3R)-2,3-Dihydroxy-1-(hydroxymethyl)heptadecyl]hexacosanamide, C26 Phytoceramide, N-(Hexacosanoyl)-phytosphingosine
InChI formel InChI=1S/C44H89NO4/c1-3-5-7-9-11-13-15-17-18-19-20-21-22-23-24-25-26-27-29-31-33-35-37-39-43(48)45-41(40-46)44(49)42(47)38-36-34-32-30-28-16-14-12-10-8-6-4-2/h41-42,44,46-47,49H,3-40H2,1-2H3,(H,45,48)/t41-,42+,44-/m0/s1
LEDER CCCCCCCCCCCCCCCCCCCCCCCCCC(=O)N[C@@H](CO)[C@H](O)[C@H](O)CCCCCCCCCCCCCC
IUPAC-namn N-[(2S,3S,4R)-1,3,4-trihydroxyoctadecan-2-yl]hexacosanamide
Molekylvikt (g/mol) 696.182
Formel vikt 695.679