Läs mer
Hexakis(1H,1H,8H-tetradecafluorooctyloxy)phosphazine, TRC
Produktkod. 30009699
Change view
Kvantitet:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009699 25 mg
30007631 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009699 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH29398825MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 186406-49-5
Molekylformel C48 H18 F84 N3 O6 P3
Molekylvikt (g/mol) 2421.46
Synonym 2,2,4,4,6,6-Hexahydro-2,2,4,4,6,6-hexakis((2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorooctyl)oxy)-1,3,5,2,4,6-triazatriphosphorine;
IUPAC-namn 2,2,4,4,6,6-hexakis(2,2,3,3,4,4,5,5,6,6,7,7,8,8-tetradecafluorooctoxy)-1,3,5-triaza-2λ5,4λ5,6λ5-triphosphacyclohexa-1,3,5-triene
LEDER FC(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)COp1(OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)F)np(OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)F)(OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)F)np(OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)F)(OCC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)F)n1
Specifikationer
Kemiskt namn eller material Hexakis(1H,1H,8H-tetradecafluorooctyloxy)phosphazine
Kvantitet 25 mg
InChI formel InChI=1S/C48H18F84N3O6P3/c49-7(50)19(73,74)31(97,98)43(121,122)37(109,110)25(85,86)13(61,62)1-136-142(137-2-14(63,64)26(87,88)38(111,112)44(123,124)32(99,100)20(75,76)8(51)52)133-143(138-3-15(65,66)27(89,90)39(113,114)45(125,126)33(101,102)21(77,78)9(53)54,139-4-16(67,68)28(91,92)40(115,116)46(127,128)34(103,104)22(79,80)10(55)56)135-144(134-142,140-5-17(69,70)29(93,94)41(117,118)47(129,130)35(105,106)23(81,82)11(57)58)141-6-18(71,72)30(95,96)42(119,120)48(131,132)36(107,108)24(83,84)12(59)60/h7-12H,1-6H2
Formel vikt 2420.907