Learn More
Hexakis(1H,1H,omegaH-perfluoroalkoxy)phosphazene, mass spectrometry standard, Thermo Scientific™
Catalog Number p-4813489
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
105809-15-2
C6H18N3P3
2120
DEBZEVJNWCNATM-MZWXYZOWSA-N
44119860
2,2,4,4,6,6-hexakis(deuteriomethyl)-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
CP1(=NP(=NP(=N1)(C)C)(C)C)C
105809-15-2
C6H18N3P3
2120
DEBZEVJNWCNATM-MZWXYZOWSA-N
44119860
2,2,4,4,6,6-hexakis(deuteriomethyl)-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
CP1(=NP(=NP(=N1)(C)C)(C)C)C
Density 1.8000g/mL
Packaging Glass bottle
Refractive Index 1.3490 to 1.3570
Quantity 250 mg
Specific Gravity 1.8
Synonym ultramark1621, ultramark 1621, hexa 2 h? methyl-1,3,5,2??,4??,6??-triazatriphosphinine
Formula Weight 2120
Chemical Name or Material Hexakis(1H, 1H, ωH-perfluoroalkoxy)phosphazene
compliance-icons
Product Identifier
  • Hexakis(1H,1H,omegaH-perfluoroalkoxy)phosphazene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.