Läs mer
Hexamethoxyphosphazine, TRC
Produktkod. 30136951
Change view
Kvantitet:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136951 250 mg
30133721 100 mg
30004685 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136951 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH293620250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 957-13-1
Molekylformel C6H18N3O6P3
Molekylvikt (g/mol) 321.14
Synonym 2,2,4,4,6,6-Hexahydro-2,2,4,4,6,6-hexamethoxy-1,3,5,2,4,6-triazatriphosphorine;
IUPAC-namn 2,2,4,4,6,6-hexamethoxy-1,3,5,2l5,4l5,6l5-triazatriphosphinine
LEDER COP1(OC)=NP(OC)(OC)=NP(OC)(OC)=N1
Specifikationer
Kemiskt namn eller material Hexamethoxyphosphazene
Anteckningar om renhetsgrad GC
Kvantitet 250 mg
InChI formel InChI=1S/C6H18N3O6P3/c1-10-16(11-2)7-17(12-3,13-4)9-18(8-16,14-5)15-6/h1-6H3
Formel vikt 321.04
Procent renhet >95%