Läs mer
Hexametyldisilazan, TRC
Specifications
Specifications
| Kemiskt namn eller material | Hexamethyldisilazane |
| CAS | 999-97-3 |
| Anteckningar om renhetsgrad | GC |
| Molekylformel | C6H19NSi2 |
| Kvantitet | 250 g |
| Synonym | 1,1,1,3,3,3-Hexamethyldisilazane, 1,1,1-Trimethyl-N-(trimethylsilyl)silanamine, A 166, A 166 (Silazane), Bis(trimethylsilyl)amine, Di(trimethylsilyl)amine, Dynasylan HMDS, HMD 3, HMDS, HMDS (silazane), HMDS 3, Hexamethyldisilazane, Hexamethyldisilylamine, LS 7150, NSC 93895, OAP, SE 31, SE 31 (silazane), SIH 6110.1, SZ 6079, TSL 8802, TSR 8802, Z 6079 |
| InChI formel | InChI=1S/C6H19NSi2/c1-8(2,3)7-9(4,5)6/h7H,1-6H3 |
| LEDER | C[Si](C)(C)N[Si](C)(C)C |
| IUPAC-namn | [dimetyl-(trimetylsilylamino)silyl]metan |
| Molekylvikt (g/mol) | 161.39 |
| Show More |