missing translation for 'onlineSavingsMsg'
Learn More

Hexamethyldisiloxane, 98+%, Thermo Scientific Chemicals

Catalog Number p-7081743
192.50 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
25 mL
100 mL
500 mL
2500 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
107-46-0
C6H18OSi2
MFCD00008265
UQEAIHBTYFGYIE-UHFFFAOYSA-N
hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide
24764
CHEBI:78002
trimethyl(trimethylsilyloxy)silane
C[Si](C)(C)O[Si](C)(C)C
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
107-46-0
C6H18OSi2
MFCD00008265
UQEAIHBTYFGYIE-UHFFFAOYSA-N
hexamethyldisiloxane, disiloxane, hexamethyl, hexamethyl disiloxane, oxybis trimethylsilane, fluka ag, hmdso, bis trimethylsilyl ether, belsil dm 0.65, bis trimethylsilyl oxide
24764
CHEBI:78002
trimethyl(trimethylsilyloxy)silane
C[Si](C)(C)O[Si](C)(C)C

Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule. It is also used to make low-k dielectric materials for the semi-conductor industries by plasma-enhanced chemical vapor deposition(PECVD) method.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Hexamethyldisiloxane is used as a solvent in organic reactions. It finds application as an international standard for 1H NMR spectroscopy for calibrating chemical shift. Further, it is used in liquid bandages to protect skin from irritation. In addition, it is used to measure tissue oxygen tension as a receptor molecule. It is also used to make low-k dielectric materials for the semi-conductor industries by plasma-enhanced chemical vapor deposition(PECVD) method.

Solubility
Slightly miscible with water.

Notes
Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, acids, bases and oxygen.
TRUSTED_SUSTAINABILITY
Melting Point -67°C
Density 0.764
Boiling Point 100°C to 101°C
Flash Point -2°C
Odor Characteristic
Linear Formula (CH3)3SiOSi(CH3)3
Refractive Index 1.377
Quantity 25 mL
UN Number UN1993
Beilstein 1736258
Solubility Information Slightly miscible with water.
Molecular Weight (g/mol) 162.38
Formula Weight 162.38
Percent Purity ≥98%
Chemical Name or Material Hexamethyldisiloxane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'