Läs mer
Hexamethylenediamine-N,N,N',N'-tetrakis(methylphosphonic Acid), TRC
Produktkod. 30403344
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30403344 250 mg
1 options
Produktkod. 30403344 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH294326250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Hexamethylenediamine-N,N,N',N'-tetrakis(methylphosphonic Acid)
CAS 23605-74-5
Molekylformel C10 H28 N2 O12 P4
Kvantitet 250 mg
Synonym Hexanediylbis[nitrilobis(methylene)]]tetrakis[phosphonic Acid], (Hexamethylenedinitrilo)tetrakis[methylenephosphonic Acid], 1,6-Hexanediaminetetrakis(methylene phosphonic Acid), Briquest 462, Dequest 2051, HDTMP, HDTMPA, Hexamethylenedinitrilotetrakis(methylphosphonic Acid), Hexanediaminetetra(methylenephosphonic Acid), N,N,N',N'-1,6-Hexamethylenediaminetetrakis(methylenephosphonic Acid), N,N,N',N'-Hexamethylenediaminetetra(methylenephosphonic Acid)
InChI formel InChI=1S/C10H28N2O12P4/c13-25(14,15)7-11(8-26(16,17)18)5-3-1-2-4-6-12(9-27(19,20)21)10-28(22,23)24/h1-10H2,(H2,13,14,15)(H2,16,17,18)(H2,19,20,21)(H2,22,23,24)
LEDER OP(=O)(O)CN(CCCCCCN(CP(=O)(O)O)CP(=O)(O)O)CP(=O)(O)O
IUPAC-namn [6-[bis(phosphonomethyl)amino]hexyl-(phosphonomethyl)amino]methylphosphonic acid
Molekylvikt (g/mol) 492.231
Formel vikt 492.059