Läs mer
Hexamethyltriamidophosphate, TRC
Produktkod. 30012827
Change view
Kvantitet:
5 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30012827 250 g
30008838 5 g
30005674 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30012827 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH294325250G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 680-31-9
Molekylformel C6 H18 N3 O P
Molekylvikt (g/mol) 179.2
Synonym HMPA, Hexamethylphosphoric triamide
IUPAC-namn N-[bis(dimethylamino)phosphoryl]-N-methylmethanamine
LEDER CN(C)P(=O)(N(C)C)N(C)C
Specifikationer
Kemiskt namn eller material Hexamethylphosphoramide (Hexametapol)
Kvantitet 250 g
InChI formel InChI=1S/C6H18N3OP/c1-7(2)11(10,8(3)4)9(5)6/h1-6H3
Formel vikt 179.1187