Läs mer
Hexane-1,6-diamine Dihydrochloride, Mikromol™
Produktkod. 30050242
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30050242 25 mg
1 options
Produktkod. 30050242 Leverantör Mikromol Leverantörsnummer MM0162.170025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Hexane-1,6-diamine Dihydrochloride
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 1,6-Diaminohexane Dihydrochloride
CAS 6055-52-3
Analyt- eller komponentnamn 1,6-Diaminohexane Dihydrochloride
Frakt skick Room Temperature
Molekylformel C6H18Cl2N2
Certifikat och efterlevnad ISO 9001
Synonym 1,6-Diamino-n-hexane, 1,6-Hexylenediamine Dihydrochloride, 80HMD, Advancure Dihydrochloride, HMDA, Hexamethylenediamine Dihydrochloride, Hexylenediamine Dihydrochloride, Hi Perm, RT Advancure HD Dihydrochloride, alpha,ω-Hexanediamine Dihydrochloride, 1,6-Hexanediamine, hydrochloride (1:2), 1,6-Hexanediamine, hydrochloride (1:2)
InChI formel InChI=1S/C6H16N2.2ClH/c7-5-3-1-2-4-6-8;;/h1-8H2;2*1H
LEDER Cl.Cl.NCCCCCCN
IUPAC-namn hexane-1,6-diamine;dihydrochloride
Molekylvikt (g/mol) 189.13
Kvantitet 25 mg
Formel vikt 188.0847 g/mol