Läs mer
Hexanedioic Acid-1-13C, TRC
Produktkod. 30324058
Change view
Kvantitet:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30324058 10 mg
30316659 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30324058 Leverantör Toronto Research Chemicals Leverantörsnummer TRCA29159810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Hexanedioic Acid-1-13C
CAS 2708283-72-9
Molekylformel 13C C5 H10 O4
Kvantitet 10 mg
Synonym Hexanedioic-1-13C acid, Adipic-1-13C acid, Hexanedioic acid-1-13C, Hexanedioic acid-13C, Adipic Acid-13C1
InChI formel InChI=1S/C6H10O4/c7-5(8)3-1-2-4-6(9)10/h1-4H2,(H,7,8)(H,9,10)/i5+1
LEDER O[13C](=O)CCCCC(=O)O
IUPAC-namn (613C)hexanedioic acid
Molekylvikt (g/mol) 147.1339
Formel vikt 147.0613