missing translation for 'onlineSavingsMsg'
Learn More

Hexanes, 95% n-Hexane, BAKER ANALYZED HPLC, J.T. Baker™

Catalog Number p-7138050
Click to view available options
Quantity:
1 L
5 L
Packaging:
EcoTainer Metal Safety Can
Glass
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
110-54-3
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
110-54-3
C6H14
86.18
MFCD02179311
VLKZOEOYAKHREP-UHFFFAOYSA-N
n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
8058
CHEBI:29021
hexane
CCCCCC

Get rapid, reproducible performance and separation results. Hexanes, 95% n-Hexane, BAKER ANALYZED HPLC, J.T. Baker™ is an application-optimized, function-tested HPLC reagent/solvent that maximizes throughput and ease-of-use without sacrificing efficiency or reproducibility.

BAKER ANALYZED HPLC Reagents are high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for research and quality control applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Recalibration or adjustments due to lot changes are minimized for optimum performance from sensitive instrumentation
  • Low backgrounds free of extraneous peaks
  • Low UV absorbance in critical ranges - assured through UV absorbance testing at a variety of points and through gradient elution testing
  • Fluorescence testing for trace impurities which would cause interference
  • Function-tested for: assay, water, minimal residue after evaporation and UV absorbance and fluorescence in critical ranges
  • Lot to lot consistency, with an actual Lot Analysis on the label
  • Innovative packaging options to assure solvent quality to the point-of-use
Melting Point -95°C
Absorbance Max. 0.2 at 210nm, Max. 0.08 at 220nm, Max. 0.008 at 254nm, Max. 0.005 at 280nm, Max. 0.005 at 350nm
Color Colorless
Linear Formula CH3(CH2)4CH3
Solubility Information 0.1g/L in water
Formula Weight 86.18g/mol
Percent Purity ≥95%
Grade For use in High Performance Liquid Chromatography
Density 0.66g/cm3
Boiling Point 69°C
Quantity 1 L
Explosion Limit 1.2 - 7.4 Vol %
Flash Point −22°C
Ignition Point 240°C
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 2 ppm
Viscosity 0.326 mPas (dynamic) / 0.50 mm2/s (kinematic)
Water Max. 0.01%
Chemical Name or Material n-Hexane
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H361f-Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'