Läs mer
Hexanoyl Glycine, TRC
Produktkod. 30136108
Change view
Kvantitet:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136108 250 mg
30135689 10 mg
30003350 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136108 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH281200250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 24003-67-6
Molekylformel C8 H15 N O3
Molekylvikt (g/mol) 173.21
Synonym Glycine, N-(1-oxohexyl)- (9CI, ACI), Glycine, N-hexanoyl- (6CI, 7CI, 8CI), N-(1-Oxohexyl)glycine (ACI), 2-Hexanamidoacetic acid, Caproylglycine, Hexanoylglycine, N-Caproylglycine, n-Hexanoylglycine, NSC 224460, N-Hexanoylglycine
IUPAC-namn 2-(hexanoylamino)acetic acid
LEDER CCCCCC(=O)NCC(=O)O
Specifikationer
Kemiskt namn eller material Hexanoyl Glycine
Anteckningar om renhetsgrad HPLC
InChI formel InChI=1S/C8H15NO3/c1-2-3-4-5-7(10)9-6-8(11)12/h2-6H2,1H3,(H,9,10)(H,11,12)
Kvantitet 250 mg
Formel vikt 173.1052
Procent renhet >95%