Läs mer
Hexanoyl-L-carnitine Chloride, TRC
Produktkod. 17963265
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17963265 1 g
17922335 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17963265 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2944401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 162067-53-0
Molekylformel C13 H26 N O4 . Cl
Molekylvikt (g/mol) 295.8
Synonym (2R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexyl)oxy]-1-propanaminium Chloride, (R)-3-Carboxy-N,N,N-trimethyl-2-[(1-oxohexyl)oxy]-1-propanaminium Chloride, (R)-3-Carboxy-2-(hexanoyloxy)-N,N,N-trimethylpropan-1-aminium Chloride
IUPAC-namn [(2R)-3-carboxy-2-hexanoyloxy-propyl]-trimethyl-ammonium;chloride
LEDER [Cl-].CCCCCC(=O)O[C@H](CC(=O)O)C[N+](C)(C)C
Specifikationer
Kemiskt namn eller material Hexanoyl-L-carnitine Chloride
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C13H25NO4.ClH/c1-5-6-7-8-13(17)18-11(9-12(15)16)10-14(2,3)4;/h11H,5-10H2,1-4H3;1H/t11-;/m1./s1
Formel vikt 295.155
Procent renhet >95%