Läs mer
Hexedine, TRC
Produktkod. 30136766
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136766 1 g
30134528 500 mg
30003687 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30136766 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2948901G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5980-31-4
Molekylformel C22 H45 N3
Molekylvikt (g/mol) 351.61
Synonym 2,6-Bis(2-ethylhexyl)-7a-methylhexahydro-1H-imidazo[1,5-c]imidazole, Hexetidine Imp. C (EP), Hexedine
IUPAC-namn 2,6-bis(2-ethylhexyl)-7a-methyl-1,3,5,7-tetrahydroimidazo[1,5-c]imidazole
LEDER CCCCC(CC)CN1CN2CN(CC(CC)CCCC)CC2(C)C1
Specifikationer
Kemiskt namn eller material 2,6-Bis(2-ethylhexyl)-7a-methylhexahydro-1H-imidazo[1,5-c]imidazole (Hexedine)
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C22H45N3/c1-6-10-12-20(8-3)14-23-16-22(5)17-24(19-25(22)18-23)15-21(9-4)13-11-7-2/h20-21H,6-19H2,1-5H3
Formel vikt 351.3613
Procent renhet >95%