Learn More
Hexyl acetate, 99%, Thermo Scientific Chemicals
Catalog Number 11984911
859.96 SEK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
142-92-7
C8H16O2
144.21
AOGQPLXWSUTHQB-UHFFFAOYSA-N
n-hexyl acetate, 1-hexyl acetate, hexyl ethanoate, acetic acid hexyl ester, n-hexyl ethanoate, acetic acid, hexyl ester, hexyl alcohol, acetate, l-hexyl acetate, fema no. 2565, natural
8908
CHEBI:87510
hexyl acetate
CCCCCCOC(=O)C
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
142-92-7
C8H16O2
144.21
AOGQPLXWSUTHQB-UHFFFAOYSA-N
n-hexyl acetate, 1-hexyl acetate, hexyl ethanoate, acetic acid hexyl ester, n-hexyl ethanoate, acetic acid, hexyl ester, hexyl alcohol, acetate, l-hexyl acetate, fema no. 2565, natural
8908
CHEBI:87510
hexyl acetate
CCCCCCOC(=O)C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Acidity 1mg KOH/g max.
Melting Point -80.0°C
Color Colorless to Yellow
Density 0.8710g/mL
Boiling Point 168.0°C to 170.0°C
Flash Point 43°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CO2C6H13
Refractive Index 1.4070 to 1.4110
Quantity 2.5 L
Beilstein 02,132
Specific Gravity 0.871
Solubility Information Solubility in water: immiscible. Other solubilities: soluble in alcohol and ether
Formula Weight 144.21
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Hexyl acetate
Show More Show Less
compliance-icons
Product Identifier
  • Hexyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement