Läs mer
Hexyl Salicylate, TRC
Produktkod. 30408537
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30408537 1 g
1 options
Produktkod. 30408537 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH2896461G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Salicylic acid-hexyl ester
CAS 6259-76-3
Molekylformel C13 H18 O3
Kvantitet 1 g
Synonym Salicylic acid, hexyl ester (6CI,7CI,8CI), 1-Hexyl salicylate, Hexyl 2-hydroxybenzoate, Hexyl salicylate, n-Hexyl salicylate
InChI formel InChI=1S/C13H18O3/c1-2-3-4-7-10-16-13(15)11-8-5-6-9-12(11)14/h5-6,8-9,14H,2-4,7,10H2,1H3
LEDER CCCCCCOC(=O)c1ccccc1O
IUPAC-namn hexyl 2-hydroxybenzoate
Molekylvikt (g/mol) 222.28
Formel vikt 222.1256