Läs mer
Hexylene Glycol, TRC
Produktkod. 30408535
Change view
Kvantitet:
50 g
Produktkod. Kvantitet
30408535 50 g
1 options
Produktkod. 30408535 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH29587050G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Hexylene Glycol
CAS 107-41-5
Molekylformel C6 H14 O2
Kvantitet 50 g
Synonym 2,4-Pentanediol, 2-methyl-, 2-Methyl-2,4-pentanediol, (±)-2-Methyl-2,4-pentanediol, 1,1,3-Trimethyltrimethylenediol, 2,4-Dihydroxy-2-methylpentane, Diolane, Hexasol, Hexylene glycol, Isohexanediol, Isol, MPD, NSC 8098, α,α,α'-Trimethyltrimethylene glycol
InChI formel InChI=1S/C6H14O2/c1-5(7)4-6(2,3)8/h5,7-8H,4H2,1-3H3
LEDER CC(O)CC(C)(C)O
IUPAC-namn 2-methylpentane-2,4-diol
Molekylvikt (g/mol) 118.17
Formel vikt 118.0994