Learn More
IBA Solutions For Life Science Hexynyl-ApG Transcription Starter
A modified RNA oligonucleotide for 5´-labeled transcripts that allows click reaction with a number of azide labels and can be used for in vitro transcription experiments.
4105.00 SEK - 17446.00 SEK
Specifications
Description Hexynyl-ApG Transcription Starter
For Use With (Application) For 5' transcript labeling, in vitro experiments
Product Type RNA oligonucleotide
View More Specs
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15633708 View Documents IBA Solutions For Life Science
5-0710-540
1 μmol 4105.00 SEK
1µmol View alternative products
Save Up to 
»
Estimated Shipment
03-10-2019
Add to basket
15643708 View Documents IBA Solutions For Life Science
5-0710-541
5 μmol 12314.00 SEK
5µmol View alternative products
Save Up to 
»
Estimated Shipment
03-10-2019
Add to basket
15653708 View Documents IBA Solutions For Life Science
5-0710-542
10 μmol 17446.00 SEK
10µmol View alternative products
Save Up to 
»
Estimated Shipment
03-10-2019
Add to basket
Description
Description
One of the main reasons for the success of IBA in custom oligonucleotide and dinucleotide synthesis is the in-house nucleic acid chemistry facility that enabless production of top quality modified oligonucleotides at competitive prices.
  • Modified dinucleotides for 5' transcript labeling: Transcription starters
  • 5'-Modified ApG can be used in in vitro transcription experiments—it is incorporated only once at the 5'-end
  • Easily introduce fluorescent markers to make transcripts visible
  • Introduce non-radioactive markers like biotin or digoxigenin to isolate transcription products
  • In principle, all of the available dyes and non-radioactive reporter groups can be introduced
Specifications
Specifications
Hexynyl-ApG Transcription Starter
For 5' transcript labeling, in vitro experiments
RNA oligonucleotide