missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HFE Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17954-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The features of hemochromatosis include cirrhosis of the liver, diabetes, hypermelanotic pigmentation of the skin, and heart failure. Since hemochromatosis is a relatively easily treated disorder if diagnosed, this is a form of preventable cancer. The HFE protein, which is defective in hereditary hemo-chromatosis, normally is expressed in crypt enterocytes of the duodenum where it has a unique, predomitly intracellular localization. In placenta, the HFE protein co-localizes with and forms a stable association with the transferrin receptor (TfR), providing a link between the HFE protein and iron transport. Immunocytochemistry shows that the HFE protein and TfR both are expressed in the crypt enterocytes. Western blots show that, as is the case in human placenta, the HFE protein in crypt enterocytes is physically associated with the TfR and with beta2-microglobulin. It is proposed that HFE has two mutually exclusive activities in cells: inhibition of uptake or inhibition of release of iron and that the balance between serum transferrin saturation and serum transferrin-receptor concentrations determines which of these functions predominates.Specifications
HFE | |
Polyclonal | |
Unconjugated | |
HFE | |
hemochromatosis, HFE, HFE1, HH, HLA H, HLAH, MVCD7 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3077 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q30201 | |
HFE | |
HFE Fusion Protein Ag12448 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |