missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HHIP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1236.20 SEK - 3313.85 SEK
Specifications
Antigen | HHIP |
---|---|
Concentration | 0.13 mg/mL |
Applications | Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838883
|
Proteintech
11654-1-AP-20UL |
20 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16828883
|
Proteintech
11654-1-AP-150UL |
150 μL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This gene encodes a protein similar to the mouse hedgehog-interacting protein, a regulatory component of the hedgehog signaling pathway. Members of the hedgehog family are evolutionarily conserved proteins which are involved in many fundamental processes in embryonic development, including anteroposterior patterns of limbs and regulation of left-right asymmetry.Specifications
HHIP | |
Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Mouse, Rat, Human | |
Q7TN16, Q96QV1 | |
15245, 291936, 64399 | |
HHIP Fusion Protein Ag2241 | |
Primary | |
-20°C | |
HHIP |
0.13 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
hedgehog interacting protein, HHIP, HIP | |
Hhip | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |