missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ HIF1A Polyclonal Antibody
GREENER_CHOICE

Produktkod. 16272321
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16272321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16272321 Leverantör Invitrogen™ Leverantörsnummer PA585494

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (90%), Rat (90%), Dog (97%), Pig (95%), Rabbit (95%), Bovine (97%). Positive Control: MCF-7, HeLa(500uM CoCl for 24hr), NIH-3T3 (500 uM CoCl2 treatment for 24 hr), HepG2 whole cell lysate/extract (1% O2 treatment for 24hr), 293T transfected HIF1A, HepG2 ( 200 uM CoCl2 treatment for 24 hr), HepG2 (500 uM CoCl2 treatment for 24 hr). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

HIF1-alpha (HIF1A) is a subunit of HIF1, which is a transcription factor found in mammalian cells cultured under reduced oxygen tension. HIF-1 is a heterodimer consisting of an alpha and beta subunit, both belonging to the basic-helix-loop-helix Per-aryl hydrocarbon receptor nuclear translocator-Sim (PAS) family of transcription factors. HIF1 functions as a transcriptional regulator of the adaptive response to hypoxia. Under hypoxic conditions, HIF-1 activates the transcription of over 40 genes, including erythropoietin, glucose transporters, glycolytic enzymes, vascular endothelial growth factor, HILPDA, and other genes whose protein products increase oxygen delivery or facilitate metabolic adaptation to hypoxia. HIF1-alpha regulates hypoxia-mediated apoptosis, cell proliferation and tumor angiogenesis. Hypoxia which induces p53 protein accumulation, directly interacts with HIF1-alpha and reduces hypoxia-induced expression of HIF1-alpha by promoting MDM2-mediated ubiquitination and proteasomal degradation under hypoxic conditions. Recent studies suggest that induction of NOX4 by HIF1-alpha contributes to maintain ROS levels after hypoxia and hypoxia-induced proliferation. In humans, it is located on the q arm of chromosome 14. The C-terminal of HIF1A binds to p300. p300/CBP-HIF complexes participate in the induction of hypoxia-responsive genes, including VEGF. Hypoxia contributes significantly to the pathophysiology of major categories of human disease, including myocardial and cerebral ischemia, cancer, pulmonary hypertension, congenital heart disease and chronic obstructive pulmonary disease.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen HIF1A
Användningsområden ChIP Assay, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Klassificering Polyclonal
Koncentration 1.47 mg/mL
Konjugera Unconjugated
Formulering PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gen HIF1A
Genaccessionsnr. O35800, Q16665, Q61221, Q9XTA5
Gene Alias AA959795; ARNT interacting protein; ARNT2; ARNT-interacting protein; Basic-helix-loop-helix-PAS protein MOP1; bHLHe78; Class E basic helix-loop-helix protein 78; HIF; hif 1; hif 1a; HIF1; HIF1 alpha; HIF-1 alpha; HIF1a; hif-1a; hif1a.L; HIF1alpha; HIF-1alpha; HIF-1-alpha; HIF1-alpha; HIF1-alpha protein; hypoxia inducible factor 1 alpha; hypoxia inducible factor 1 alpha subunit; hypoxia inducible factor 1 alpha subunit L homeolog; hypoxia inducible factor 1 subunit alpha; hypoxia inducible factor 1 subunit alpha L homeolog; hypoxia inducible factor 1, alpha subunit; hypoxia inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia inducible factor-1 alpha; hypoxia-inducible factor 1 alpha; hypoxia-inducible factor 1 alpha isoform I.3; hypoxia-inducible factor 1 alpha subunit; hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor); hypoxia-inducible factor 1, alpha subunit (basic helix-loop-helix transcription factor) L homeolog; hypoxia-inducible factor 1-alpha; hypoxia-inducible factor-1 alpha; hypoxia-inducible factor1alpha; member of PAS protein 1; member of PAS superfamily 1; MOP1; PAS domain-containing protein 8; PASD8; XELAEV_18039792mg; XELAEV_18041137mg; xhif1a
Gensymboler HIF1A
Värd art Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human HIF1 alpha. The exact sequence is proprietary.
Reningsmetod Antigen affinity chromatography
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 100009579, 15251, 281814, 29560, 3091
Målarter Human, Mouse, Rat, Rabbit, Bovine
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.