missing translation for 'onlineSavingsMsg'
Learn More

HIF1AN Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 10646-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16887153

  • 3313.85 SEK / 150µL
Estimated Shipment: 30-11-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Factor Inhibiting HIF1 (FIH) protein facilitates repression of HIF-1 transcriptional activity by binding to VHL, which acts as a transcriptional corepressor. VHL inhibits HIF-1 alpha transactivation function by recruiting histone deacetylases. Involvement of VHL in association with FIH provides a unifying mechanism for the modulation of HIF-1 alpha protein stabilization and transcriptional activation in response to changes in cellular oxygen concentration.
Specifications

Specifications

HIF1AN
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BLR9, Q9NWT6
Factor inhibiting HIF 1, FIH 1, FIH1, HIF1AN
Rabbit
IgG
150 μL
-20°C
Primary
309434, 319594, 55662
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HIF1AN
Liquid
Hif1an
HIF1AN Fusion Protein Ag0997
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Videos
SDS
Documents
Certificates
Special Offers

Special Offers