Learn More
HIGD1B Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25367-1-AP-20UL
Additional Details : Weight : 0.00010kg
Specifications
HIGD1B | |
Polyclonal | |
Unconjugated | |
HIGD1B | |
CLST11240, CLST11240 15, HIG1 domain family member 1B, HIGD1B, Protein CLST 11240 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51751 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9P298 | |
HIGD1B | |
HIGD1B Fusion Protein Ag21871 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |