Learn More
Contec™ High Duster XF with Hand Grip
Product Code. 30274784 Shop All Contec Products
Change view
Click to view available options
Length (English):
24.25 in.
Unit Size:
Each
Product Code. Length (English) Unit Size
30274784 24.25 in. Each
1 options
Product Code. 30274784 Supplier Contec™ Supplier No. MGMP0008
  • ONLINE EXCLUSIVE PRICE
    887.00 SEK
    683.81 SEK / Paket med 1
    Save 203.19 SEK
    22% Av
Beräknad leverans: 11-01-2027
The High Duster XF makes cleaning hard-to-reach spaces easier when paired with MicroGenesis Microfiber High Duster Covers. The flexible body of the high duster tool bends to hug surface contours and reduces operator strain.
  • Flexible frame reaches areas where typical dusters cannot
  • Reduces cross contamination risk when paired with a Microfiber High Duster Cover
TRUSTED_SUSTAINABILITY
Specifications
Product Type Duster Tool
Flammable No
Length (English) 24.25 in.
Length (Metric) 62 cm