Learn More
Merck High Output Pump
High Output Pump features a piston-driven design for greater power.
Brand:  Merck WP6210060
Product Code. 12444257
  • 22610.00 SEK / Each
Only null left
Estimated Shipment: 06-08-2025
Description
  • High flow rates decrease filtration process time
  • Optional chemical-resistant internal components for compatibility with corrosive vapors
  • Portable design allows for easy sharing between workstations
TRUSTED_SUSTAINABILITY
Specifications
Specifications
70 cm of 6.35 mm tubing and a Millex™-FA50 filter
UL-listed and CE-marked
100 V 50/60 Hz
Vacuum Pump
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Merck High Output Pump > Electrical Requirements: 100 V, 50/60 Hz