Läs mer
Scotch-Brite™ Högpresterande mikrofiberservetter 2010
Färg:
Blått
Grön
Gult
Rött
Vitt
Beskrivning
- High performance microfibre wipe offering highly efficient, and lint free cleaning on virtually any surface.
- High quality cleaning results even when used with or without water, and with or without detergent
- Extremely durable, and can resist up to 500 wash cycles
- Machine washable up to 95°C
Specifikationer
Specifikationer
| Färg | Rött |
| Mått (L x B) | 320 x 360 mm |
| Förpackningsmängd | 10/Pk. |