Learn More
HINT1 Mouse anti-Human, Unconjugated, Clone: 1G3E4, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67583-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840296
  • 1656.73 SEK / 20µL
Estimated Shipment: 14-05-2024
to see stock.
Description
Description
The HINT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HINT1. This antibody recognizes human antigen. The HINT1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
HINT1
Monoclonal
1500 μg/mL
Mouse
Primary
Human
Western Blot, Immunohistochemistry, ELISA
1G3E4
Unconjugated
20 μL
3094
IgG2a