Learn More
HINT1 Mouse anti-Human, Unconjugated, Clone: 1G3E4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67583-1-IG-20UL
Additional Details : Weight : 0.00010kg
Description
The HINT1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human HINT1. This antibody recognizes human antigen. The HINT1 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
HINT1 | |
Monoclonal | |
1500 μg/mL | |
Mouse | |
Primary | |
Human |
Western Blot, Immunohistochemistry, ELISA | |
1G3E4 | |
Unconjugated | |
20 μL | |
3094 | |
IgG2a |