missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Hippocalcin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA519535
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Heat mediated antigen retrieval recommended prior to tissue staining. This antibody is predicted to react with chicken and cow based on sequence homology.
The protein encoded by this gene is a member of neuron-specific calcium-binding proteins family found in the retina and brain. This protein is associated with the plasma membrane. It has similarities to proteins located in the photoreceptor cells that regulate photosignal transduction in a calcium-sensitive manner. This protein displays recoverin activity and a calcium-dependent inhibition of rhodopsin kinase. It is identical to the rat and mouse hippocalcin proteins and thought to play an important role in neurons of the central nervous system in a number of species.
Specifikationer
| Hippocalcin | |
| Polyclonal | |
| Unconjugated | |
| HPCA | |
| BDR2; calcium-binding protein; calcium-binding protein BDR-2; DYT2; hippocalcin; Hpca; hypothetical protein MGC73126; neuron-specific calcium-binding protein hippocalcin; P23K; zgc:73126 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15444, 29177, 3208, 393696 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry, Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide; pH 7.4 | |
| P84074, P84075, P84076 | |
| HPCA | |
| Synthetic peptide conjugated to KLH derived from within residues 150 to the C-terminus of Rat Hippocalcin. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat, Zebrafish | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur