Läs mer
Hirsutidin Chloride, TRC
Produktkod. 30404606
Change view
Kvantitet:
0.5 mg
2.5 mg
Förpackningsstorlek:
0.5mg
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30404606 0.5 mg 0.5mg
30407883 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30404606 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH3568000.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 4092-66-4
Molekylformel C18 H17 O7 . Cl
Molekylvikt (g/mol) 380.776
Synonym 3,5-Dihydroxy-2-(4-hydroxy-3,5-dimethoxyphenyl)-7-methoxy-1-benzopyrylium Chloride, 3,4',5-Trihydroxy-3',5',7-trimethoxyflavylium Chloride, Hirsutidin, Hirsutinidol Chloride
IUPAC-namn 2-(4-hydroxy-3,5-dimethoxyphenyl)-7-methoxychromenylium-3,5-diol;chloride
LEDER [Cl-].COc1cc(O)c2cc(O)c([o+]c2c1)c3cc(OC)c(O)c(OC)c3
Specifikationer
Kemiskt namn eller material Hirsutidin Chloride
Kvantitet 0.5 mg
InChI formel InChI=1S/C18H16O7.ClH/c1-22-10-6-12(19)11-8-13(20)18(25-14(11)7-10)9-4-15(23-2)17(21)16(5-9)24-3;/h4-8H,1-3H3,(H2-,19,20,21);1H
Formel vikt 380.066