missing translation for 'onlineSavingsMsg'
Learn More
Learn More
His Tag Antibody (3D5), Alexa Fluor™ 350, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP2-81021AF350
This item is not returnable.
View return policy
Description
His Tag Monoclonal antibody specifically detects His Tag in Epitope Tag samples. It is validated for Western Blot, ELISA.Specifications
His Tag | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Human | |
Protein A purified | |
RUO | |
Primary | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
3D5 | |
Western Blot, ELISA | |
6 His epitope tag, 6X His, 6X-His, H, HHHHHH epitope tag, HHHHHH tag, HIS, His tag, Poly-histidine | |
Purified his tagged Fab fragment. | |
0.1 mL | |
Cellular Markers, Epitope Tags | |
This antibody binds to carboxy-terminal oligo-histidine tags (6x His tags) of fusion proteins. It also has affinity for internal His-tags but no affinity for N-terminal His-tags. | |
Purified | |
Epitope Tag |