missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIST1H2AC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | HIST1H2AC |
---|---|
Concentration | 0.35 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16835364
|
Proteintech
15953-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16825364
|
Proteintech
15953-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 02-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
HIST1H2AC | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
C0HKE2, Q93077 | |
306962, 319164, 8334 | |
HIST1H2AC Fusion Protein Ag8663 | |
Primary | |
-20°C | |
HIST1H2AC |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
H2A/l, H2AFL, HIST1H2AC, histone cluster 1, H2ac, Histone H2A type 1 C, Histone H2A/l | |
Hist1h2ac | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |