missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ HIST1H2BC Recombinant Protein
Brand: Abnova™ H00008347-P01.25ug
Additional Details : Weight : 0.00010kg
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH09612 | |
Solution | |
8347 | |
HIST1H2BC (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HIST1H2BC | |
Human | |
Yes |
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein) | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
39.6 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
MPEPAKSAPAPKKGSKKAVTKAQKKDGKKRKRSRKESYSVYVYKVLKQVHPDTGISSKAMGIMNSFVNDIFERIAGEASRLAHYNKRSTITSREIQTAVRLLLPGELAKHAVSEGTKAVTKYTSSK | |
H2B.1/H2B/l/H2BFL/MGC104246/dJ221C16.3 | |
HIST1H2BC | |
Wheat Germ (in vitro) | |
GST |